Learn More
Fine Science Tools Sample Corers, 15 Gauge, 8.5 cm
SDP
Catalog No. NC9128328
Encompass
Product: Sample Corer - 15G/ Gauge : 15/ ID : 1mm/ OD : 1.8mm/ Alloy : Stainless Steel/ Length : 8.5cm/ Unit : Each
Catalog No. NC9128328 Supplier Fine Science Tools Supplier No. 1803501
Product: Sample Corer - 15G/ Gauge : 15/ ID : 1mm/ OD : 1.8mm/ Alloy : Stainless Steel/ Length : 8.5cm/ Unit : Each