Learn More
Premier Lab Supply 32mm XRF Sample Cups – PN# SC-4231
SDP
Catalog No. NC0447207
Encompass_Preferred
32mm Double-Open Ended XRF Sample Cup with Cup & 2 Rings (PN# SC-4231) – Suitable for Universal – OD: 32mm; ID: 24mm; Height: 24mm; Volume: 12mL – 100 per bag
Catalog No. NC0447207 Supplier Premier Lab Supply Supplier No. SC4231
32mm Double-Open Ended XRF Sample Cup with Cup & 2 Rings (PN# SC-4231) – Suitable for Universal – OD: 32mm; ID: 24mm; Height: 24mm; Volume: 12mL – 100 per bag