Learn More
Brookfield Engineer Labs Sample Cup Assy. RTD E-Gap 316SS with Cable Brookfield Ametek CPA-44PYZ
Catalog No. NC0426976
Encompass
Wells Brookfield Ametek Sample Cup Assembly,Embedded RTD Temperature Sensor, Electronic Gap Setting, 316SS, with Connecting Cable SC4-61Y to Display Temperature on Compatible Viscometer.
Catalog No. NC0426976 Supplier Brookfield Engineer Labs Supplier No. CPA44PYZ
Wells Brookfield Ametek Sample Cup Assembly,Embedded RTD Temperature Sensor, Electronic Gap Setting, 316SS, with Connecting Cable SC4-61Y to Display Temperature on Compatible Viscometer.