Learn More
Alkali Scientific Sample Cup with Attached Snap Cap for Roche Cobas Mira, Bio, Fara & Horiba ABX Analyzers, 1,500/Pk
SDP
Catalog No. 5000662376
Encompass_Preferred
These clear sample cups with caps are engineered for precise performance on Roche Cobas Mira, Bio, Fara and Horiba ABX analyzers. Manufactured using high quality, laboratory grade materials, they provide excellent sample visibility, accurate volume handling and secure sealing to prevent leakage, evaporation or contamination. Each cup is precision molded to fit seamlessly into analyzer sample trays, ensuring reliable compatibility and smooth workflow operation. Produced in ISO compliant facilities and suitable for use in CLIA certified laboratories, these sample cups meet the quality, consistency and traceability standards required for clinical diagnostics and research environments. The secure snap cap maintains sample integrity during handling, transport and storage. These cups are a cost effective alternative to OEM consumables without compromising accuracy or performance. Available in other colors upon request to accommodate laboratory identification systems or specific workflow.
Catalog No. 50-006-62376 Supplier Alkali Scientific Supplier No. AMTROCHE
These clear sample cups with caps are engineered for precise performance on Roche Cobas Mira, Bio, Fara and Horiba ABX analyzers. Manufactured using high quality, laboratory grade materials, they provide excellent sample visibility, accurate volume handling and secure sealing to prevent leakage, evaporation or contamination. Each cup is precision molded to fit seamlessly into analyzer sample trays, ensuring reliable compatibility and smooth workflow operation. Produced in ISO compliant facilities and suitable for use in CLIA certified laboratories, these sample cups meet the quality, consistency and traceability standards required for clinical diagnostics and research environments. The secure snap cap maintains sample integrity during handling, transport and storage. These cups are a cost effective alternative to OEM consumables without compromising accuracy or performance. Available in other colors upon request to accommodate laboratory identification systems or specific workflow.