Learn More
Heathrow Scientific Sample Cup Containers, 20 mL
Description
- Disposable sample cup containers with lids for consistent sample preparation and repeatable water activity testing
- Pack of 10 sample cups with lids
- To use for sampling: Remove lid and place sample in cup
- Thread cup to water activity meter detachable sensor unit and connect to unit to begin measurement
- Use lid for sample storage
Specifications
Specifications
| Product Type | Sample Holder |
| For Use With (Equipment) | Water Activity Meter |
| Quantity | 10 Pk. |