Learn More
Chemplex Industries SAMPLE CUPS 31.0MM DIA. DBL OP
Catalog No. 50249397
Encompass_Preferred
CHEMPLEX XRF SAMPLE CUPS, 31 Dia., mm, Double open ends, Snap-On Rings have integrated thin-film and Microporous Film trimmer
Catalog No. 50-249-397 Supplier Chemplex Industries Supplier No. 1530SE
CHEMPLEX XRF SAMPLE CUPS, 31 Dia., mm, Double open ends, Snap-On Rings have integrated thin-film and Microporous Film trimmer