Learn More
Chemplex Industries SAMPLE CUPS 31.0MM DIA. SGL OP
Catalog No. 50249393
Encompass_Preferred
CHEMPLEX XRF SAMPLE CUPS, 31 Dia., mm, Single open end, Snap-On Rings have integrated thinfilm trimmer
Catalog No. 50-249-393 Supplier Chemplex Industries Supplier No. 1430SE
CHEMPLEX XRF SAMPLE CUPS, 31 Dia., mm, Single open end, Snap-On Rings have integrated thinfilm trimmer