Learn More
Chemplex Industries SAMPLE CUPS 31.5MM DIA. DBL OP
Catalog No. 50249412
Encompass_Preferred
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 31.5 Dia., mm, Double open ended, Internal Overflow Reservoir
Catalog No. 50-249-412 Supplier Chemplex Industries Supplier No. 2132S
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 31.5 Dia., mm, Double open ended, Internal Overflow Reservoir