Learn More
Chemplex Industries SAMPLE CUPS 39.4MM DIA. SGL OP
Catalog No. 50249395
Encompass_Preferred
CHEMPLEX XRF SAMPLE CUPS, 39.4 Dia., mm, Single open end for Replacement of Oxford 54-LX6922 Sample Cups
Catalog No. 50-249-395 Supplier Chemplex Industries Supplier No. 1440L
CHEMPLEX XRF SAMPLE CUPS, 39.4 Dia., mm, Single open end for Replacement of Oxford 54-LX6922 Sample Cups