Learn More
Chemplex Industries SAMPLE CUPS 40.1MM DIA. DBL OP
Catalog No. 50249414
Encompass_Preferred
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 40.1 Dia., mm, Double open ended, 43.2mm Max O.D., Internal Overflow Reservoir
Catalog No. 50-249-414 Supplier Chemplex Industries Supplier No. 2140
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 40.1 Dia., mm, Double open ended, 43.2mm Max O.D., Internal Overflow Reservoir