Learn More
Chemplex Industries SpectroSulfur Analyzer Low Profile XRF Sample Cup
Catalog No. 50249405
Encompass_Preferred
CHEMPLEX XRF SAMPLE CUPS, 42.9 Dia., mm, .77 (19.6mm) Height, Single open end, SpectroSulfur(TM) Analyzer Sample Cups
Catalog No. 50-249-405 Supplier Chemplex Industries Supplier No. 1850
CHEMPLEX XRF SAMPLE CUPS, 42.9 Dia., mm, .77 (19.6mm) Height, Single open end, SpectroSulfur(TM) Analyzer Sample Cups