Learn More
Chemplex Industries SAMPLE CUPS 43.4MM DIA. DBL OP
Catalog No. 50249419
Encompass_Preferred
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 43.4 Dia., mm, Double open ended, Internal Overflow Reservoir
Catalog No. 50-249-419 Supplier Chemplex Industries Supplier No. 2195
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 43.4 Dia., mm, Double open ended, Internal Overflow Reservoir