Learn More
Chemplex Industries SAMPLE CUPS 44.7MM DIA. DBL OP
Catalog No. 50249417
Encompass_Preferred
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 44.7 Dia., mm, Double open ended, with Cap, Internal Overflow Reservoir
Catalog No. 50-249-417 Supplier Chemplex Industries Supplier No. 2145
CHEMPLEX XRF SAMPLE CUPS, SpectroCup(TM), 44.7 Dia., mm, Double open ended, with Cap, Internal Overflow Reservoir