Learn More
TA Instruments Sample Cutting Kit, Includes Tools for Preparing Samples for Consistent Results, Pliers, Style Cutter, 4 mm Hole Punch, Hammer, Ensures Flat Surfaces for Tzero Pans
Catalog No. 5000580716 Shop All TA Instruments Products
Encompass_Preferred
Waters TA Instruments Sample Cutting Kit includes tools for proper preparation of samples to ensure repeatability and reliability. It features a pliers style cutter for clean flat surfaces, a 4 mm hole punch and hammer for consistent sample sizing, especially for Tzero pans. This kit ensures optimal contact and improved measurement reliability.
Catalog No. 50-005-80716 Supplier TA Instruments Supplier No. 973727.901
Waters TA Instruments Sample Cutting Kit includes tools for proper preparation of samples to ensure repeatability and reliability. It features a pliers style cutter for clean flat surfaces, a 4 mm hole punch and hammer for consistent sample sizing, especially for Tzero pans. This kit ensures optimal contact and improved measurement reliability.