Learn More
New Jersey Fluid Systems LLC SAMPLE CYLINDER
Catalog No. NC0072215
Encompass
304L SS Double-Ended DOT-Compliant Sample Cylinder, 1/4 in. FNPT, 1000 cm3, 1800 psig (124 bar)
Catalog No. NC0072215 Supplier New Jersey Fluid Systems LLC Supplier No. 304LHDF41000
304L SS Double-Ended DOT-Compliant Sample Cylinder, 1/4 in. FNPT, 1000 cm3, 1800 psig (124 bar)