Learn More
Cygnus Technologies SAMPLE DILUENT BUFFER 100 ML
Catalog No. NC9689565
Encompass
Used to dilute out high levels of Host Cell Proteins in samples.
Catalog No. NC9689565 Supplier Cygnus Technologies Supplier No. I028100
Used to dilute out high levels of Host Cell Proteins in samples.