Learn More
Perkin Elmer US LLC Sample Disk Holder for Spotlight Systems
Catalog No. 502112661
Encompass_Preferred
The Sample Disk Holder is made of an aluminum alloy and accommodates up to 3 sampling discs, 13 mm in diameter. It is 76 (L) x 25 (W) x 3 (H) mm and sits on the sample stage of PerkinElmer's Spotlight systems. With this part number, you can order a replacement or spare holder.
Catalog No. 50-211-2661 Supplier Perkin Elmer US LLC Supplier No. L1861634
The Sample Disk Holder is made of an aluminum alloy and accommodates up to 3 sampling discs, 13 mm in diameter. It is 76 (L) x 25 (W) x 3 (H) mm and sits on the sample stage of PerkinElmer's Spotlight systems. With this part number, you can order a replacement or spare holder.