Learn More
Fritsch Milling And Sizing Inc Sample Glass 2 Litres Wide Mouth Bottle Gl 80
SDP
Catalog No. 501534711
Encompass
Is recommended, if a larger sample collection as 500 ml is required, because the sample exhauster with cyclone separator is delivered with a 500 ml glass bottle as standard. Also 1 litre and 5 litres of sample glasses are available.
Please order the adapter additionally - article no. 19.5790.00 - to attach the 2 litres sample glass.
Catalog No. 50-153-4711 Supplier Fritsch Milling And Sizing Inc Supplier No. 83.3260.00
Is recommended, if a larger sample collection as 500 ml is required, because the sample exhauster with cyclone separator is delivered with a 500 ml glass bottle as standard. Also 1 litre and 5 litres of sample glasses are available.
Please order the adapter additionally - article no. 19.5790.00 - to attach the 2 litres sample glass.