Learn More
Bioscience International Inc Bioscience SAS Petri Style Sampling Head, Sterile, Disposable, No Autoclaving Required, 40 per Box
SDP
Catalog No. NC2327501
Encompass
The Bioscience International SAS Petri Style Sampling Head is a sterile, disposable air sampling head designed for efficient microbial air sampling in cleanrooms and controlled environments. It integrates seamlessly with various SAS air samplers, eliminating the need for autoclaving, thus enhancing workflow. Each unit is triple-wrapped for sterility, compatible with standard Petri dishes, and has a shelf life of 5 years. It is ideal for applications in pharmaceuticals, electronics, and food technology.
Catalog No. NC2327501 Supplier Bioscience International Inc Supplier No. 23236
Add to Cart
Edge
Add to Cart
The Bioscience International SAS Petri Style Sampling Head is a sterile, disposable air sampling head designed for efficient microbial air sampling in cleanrooms and controlled environments. It integrates seamlessly with various SAS air samplers, eliminating the need for autoclaving, thus enhancing workflow. Each unit is triple-wrapped for sterility, compatible with standard Petri dishes, and has a shelf life of 5 years. It is ideal for applications in pharmaceuticals, electronics, and food technology.