Learn More
Electron Microscopy Sciences Tripod Polisher Sample Holder
SDP
Catalog No. 50980871
Encompass
Mounted in the center of the Tripod Polisher Base in place of the L-Bracket and is used for parallel polishing. For use with Tripod Polisher Model 590.
Catalog No. 50-980-871 Supplier Electron Microscopy Sciences Supplier No. 5011635
Mounted in the center of the Tripod Polisher Base in place of the L-Bracket and is used for parallel polishing. For use with Tripod Polisher Model 590.