Learn More
Genevac Inc Genevac 10-5038 Vial Holder, Solid Aluminum, 24 Vial Capacity, Optimized Heat Transfer
Catalog No. NC3312536
Encompass
The Genevac Sample Holder is designed for efficient and safe sample processing in centrifugal evaporators. Made from hard-anodised aluminum, it minimizes glassware breakage and provides protection against organic solvents and strong acids. With the ability to hold 24 vials, it features optimized heat transfer for superior evaporation performance, making it a reliable choice for laboratories looking for durability and performance in sample handling.
Catalog No. NC3312536 Supplier Genevac Inc Supplier No. 105038
The Genevac Sample Holder is designed for efficient and safe sample processing in centrifugal evaporators. Made from hard-anodised aluminum, it minimizes glassware breakage and provides protection against organic solvents and strong acids. With the ability to hold 24 vials, it features optimized heat transfer for superior evaporation performance, making it a reliable choice for laboratories looking for durability and performance in sample handling.