Learn More
Perkin Elmer US LLC Sample Holder Cover Reforming Tool
Catalog No. 502113481
Encompass_Preferred
DSC performance is enhanced with sample holder covers that are formed properly and not degraded by extended use. This tool allows the analyst to reform the platinum covers to their orginial dimensions.
Catalog No. 50-211-3481 Supplier Perkin Elmer US LLC Supplier No. 03190030
DSC performance is enhanced with sample holder covers that are formed properly and not degraded by extended use. This tool allows the analyst to reform the platinum covers to their orginial dimensions.