Learn More
Perkin Elmer US LLC Ceramic Sample Cup Holder
Catalog No. 509044757
Encompass_Preferred
The Ceramic Sample Cup Holder is a sample handling tool that comes standard in the TGA 4000 Non-AutoSampler Accessory Kit (N5200046). This tool can also be used when working with ceramic sample holders and the following analyzers: TGA 6/TGA 4000/STA 6000. Qty. 1
Catalog No. 50-904-4757 Supplier Perkin Elmer US LLC Supplier No. N5203020
The Ceramic Sample Cup Holder is a sample handling tool that comes standard in the TGA 4000 Non-AutoSampler Accessory Kit (N5200046). This tool can also be used when working with ceramic sample holders and the following analyzers: TGA 6/TGA 4000/STA 6000. Qty. 1