Learn More
Mettler Toledo Sample Injection Adapter NS14
Catalog No. 01672602 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Injection Adapter
Catalog No. Type
01-672-602 Injection Adapter
1 options
Catalog No. 01-672-602 Supplier Mettler Toledo Supplier No. 30988538
Add to Cart
Add to Cart
Designed to allow samples to be injected into the titration vessel/system, facilitating moisture analysis of liquid or dissolved samples
  • The Sample Injection Adapter NS14 is an accessory from Mettler Toledo, intended for use with their coulometric Karl-Fischer titrators.
  • The adapter is used in conjunction with the titrator and the associated titration vessel/cell.
  • It enables controlled injection of a sample into the titration medium i.e. helps introduce the test sample (liquid or solution) safely and accurately without compromising the titration system.
  • This is essential for moisture analysis in labs where coulometric KF titration is used.
TRUSTED_SUSTAINABILITY
Specifications
Type Injection Adapter