Learn More
Trajan Scientific SAMPLE INJECTOR 10R-GT
SDP
Catalog No. 502530400
Encompass_Preferred
SGE Gastight Syringes: Removable-Needle Models, Diameter Outer: 0.25 in., 6.5 mm, Needle Gauge: 26 G, Volume: 10 uL, Needle Point Style: Removable Bevel, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer Needle: 0.01 in., 0.47 mm
Catalog No. 50-253-0400 Supplier Trajan Scientific Supplier No. 002250
SGE Gastight Syringes: Removable-Needle Models, Diameter Outer: 0.25 in., 6.5 mm, Needle Gauge: 26 G, Volume: 10 uL, Needle Point Style: Removable Bevel, Length Needle: 50 mm, 1.96 in., Accuracy: +/-1%, Connection Type: Removable, Diameter Outer Needle: 0.01 in., 0.47 mm