Learn More
Waters Corp Waters Tubing Assembly, Sample Inlet, SS 316, 1/16 in. OD x 0.009 in. ID
Catalog No. 50792620 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Tubing Assembly, Sample Inlet, SS 316, 1/16 in. OD x 0.009 in. ID
Catalog No. 50-792-620 Supplier Waters Corp Supplier No. 700001710
Waters Tubing Assembly, Sample Inlet, SS 316, 1/16 in. OD x 0.009 in. ID