Learn More
BURKLE INC Sample insert, 0.02 oz. (0,5 ml), Novartos Multi
Catalog No. 5000025782
Encompass_Preferred
Novartos Multi sample inserts are precisely fitting, easily interchangeable sample inserts for the Novartos Multi pharmaceutical sampler. Different volumes can be selected and combined. The blank insert (0.0 ml) can be used to prevent sampling at a specific point.
Catalog No. 50-000-25782 Supplier BURKLE INC Supplier No. 53580005
Novartos Multi sample inserts are precisely fitting, easily interchangeable sample inserts for the Novartos Multi pharmaceutical sampler. Different volumes can be selected and combined. The blank insert (0.0 ml) can be used to prevent sampling at a specific point.