Learn More
BURKLE INC Sample insert, 0.02 oz. (0,5 ml), Novartos Uno
Catalog No. 5000025788
Encompass_Preferred
Novartos Uno sample inserts are precisely fitting, easily exchangeable sample inserts for the Novartos Uno pharmaceutical sampler. Different volumes can be selected and combined.
The blank insert (0.0 ml) can be used to prevent sampling at a specific point.
Catalog No. 50-000-25788 Supplier BURKLE INC Supplier No. 53580105
Novartos Uno sample inserts are precisely fitting, easily exchangeable sample inserts for the Novartos Uno pharmaceutical sampler. Different volumes can be selected and combined.
The blank insert (0.0 ml) can be used to prevent sampling at a specific point.