Learn More
BURKLE INC Sample insert, 0.10 oz. (3 ml), Novartos Uno
Supplier:  BURKLE INC 53580130
Encompass_Preferred
Novartos Uno sample inserts are precisely fitting, easily exchangeable sample inserts for the Novartos Uno pharmaceutical sampler. Different volumes can be selected and combined.
The blank insert (0.0 ml) can be used to prevent sampling at a specific point.
Catalog No. 50-000-25582