Learn More
BURKLE INC Sample insert, 0.0 oz. (0,0 ml), Novartos Multi
Catalog No. 5000026058
Encompass_Preferred
Novartos Multi sample inserts are precisely fitting, easily interchangeable sample inserts for the Novartos Multi pharmaceutical sampler. Different volumes can be selected and combined. The blank insert (0.0 ml) can be used to prevent sampling at a specific point.
Catalog No. 50-000-26058 Supplier BURKLE INC Supplier No. 53580000
Novartos Multi sample inserts are precisely fitting, easily interchangeable sample inserts for the Novartos Multi pharmaceutical sampler. Different volumes can be selected and combined. The blank insert (0.0 ml) can be used to prevent sampling at a specific point.