Learn More
BURKLE INC Sample insert, 0.03 oz. (1 ml), Novartos Uno
Catalog No. 5000025595
Encompass_Preferred
Novartos Uno sample inserts are precisely fitting, easily exchangeable sample inserts for the Novartos Uno pharmaceutical sampler. Different volumes can be selected and combined.
The blank insert (0.0 ml) can be used to prevent sampling at a specific point.
Catalog No. 50-000-25595 Supplier BURKLE INC Supplier No. 53580110
Novartos Uno sample inserts are precisely fitting, easily exchangeable sample inserts for the Novartos Uno pharmaceutical sampler. Different volumes can be selected and combined.
The blank insert (0.0 ml) can be used to prevent sampling at a specific point.