Learn More
Chemglass Life Sciences 9" Sample Line Assembly, L/S Platinum Cured Silicone Pump Tubing, 1/8" ID x 1/4" OD Size 16
SDP
Catalog No. 501294422
Encompass
Reduces set up time and minimizes labor cost.Sample line assembly is individually bagged and ready to use.Swabable valvecan be used for sampling bioreactors.Manufactured with platinum cured siliconetubing.For sterile applications the Sampler can be safely autoclaved.
Catalog No. 50-129-4422 Supplier Chemglass Life Sciences Supplier No. CLS139013
Reduces set up time and minimizes labor cost.Sample line assembly is individually bagged and ready to use.Swabable valvecan be used for sampling bioreactors.Manufactured with platinum cured siliconetubing.For sterile applications the Sampler can be safely autoclaved.