Learn More
Perkin Elmer US LLC SAMPLE LOOP 0.5 ML
Catalog No. NC2043463
Encompass_Preferred
500 µL Sample Loop, 0.8 mm ID, connect to ports #1 and #4. Connects to ports, #1 and #4 SC-FAST high-flow PFA crimp free SC-FAST loops provide low resistance for fast loading and rinse out. Multipurpose PFA FAST loops for high-flow valves. Recommended for use with 0.8 mm i.d. (blue) probe. 1.6 mm o.d., 1/4-28 fittings.
Catalog No. NC2043463 Supplier Perkin Elmer US LLC Supplier No. N0777289
500 µL Sample Loop, 0.8 mm ID, connect to ports #1 and #4. Connects to ports, #1 and #4 SC-FAST high-flow PFA crimp free SC-FAST loops provide low resistance for fast loading and rinse out. Multipurpose PFA FAST loops for high-flow valves. Recommended for use with 0.8 mm i.d. (blue) probe. 1.6 mm o.d., 1/4-28 fittings.