Learn More
Thermo Scientific Sample Loop, NanoViper
Supplier:  Thermo Scientific 6826.2405
Encompass
5μL Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000(T)PL RS, maximum pressure: 14503 psi (1000 bar)
Catalog No. 50-134-8362