Learn More
Thermo Scientific Sample Loops for UltiMate WPS-3000T PL RS Pulled Loop Autosamplers | | 1 EA
Catalog No. 501348364 Shop All Thermo Scientific Products
Encompass
Incorporates Thermo Scientific Dionex nanoViper Fingertight Fittings, Eliminates dead volumes, Ensures easy, perfect connections, Withstands pressures up to 1000 bar, Compatible with third-party valves and unions
This item is not returnable. View return policy
Catalog No. 50-134-8364 Supplier Thermo Scientific Supplier No. 6826.2412
Incorporates Thermo Scientific Dionex nanoViper Fingertight Fittings, Eliminates dead volumes, Ensures easy, perfect connections, Withstands pressures up to 1000 bar, Compatible with third-party valves and unions