Learn More
Trajan Scientific VICI Valco Sample Loop Valco 100l Stainless Steel (VAL.SL100CW)
Catalog No. 50635017
Encompass
VICI Valco 1/16" Stainless Steel Loops Includes Two Stainless Nuts and Two Stainless ferrµles. VICI Valco Part Number: SL100CW (NEW LPP PN: VAL.SL100CW). Thermo Part Number: CH-952460. Made in the USA.
Catalog No. 50-635-017 Supplier Trajan Scientific Supplier No. PAL.SL100CW
VICI Valco 1/16" Stainless Steel Loops Includes Two Stainless Nuts and Two Stainless ferrµles. VICI Valco Part Number: SL100CW (NEW LPP PN: VAL.SL100CW). Thermo Part Number: CH-952460. Made in the USA.