Learn More
Waters Corp Waters Loop, 10 μL, HPLC System, for Rheodyne Injectors
Catalog No. 50793489 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample loop is designed for high-performance liquid chromatography (HPLC) systems. It features a 10 μL volume and is compatible with Rheodyne 7010 and 7125 injectors.
  • Designed for HPLC systems.
  • Features a 10 μL volume.
  • Compatible with Rheodyne 7010 and 7125 injectors.
  • Supplied as 1 unit per package.
  • Replaced as required for optimal performance.
Catalog No. 50-793-489 Supplier Waters Corp Supplier No. WAT007041
This sample loop is designed for high-performance liquid chromatography (HPLC) systems. It features a 10 μL volume and is compatible with Rheodyne 7010 and 7125 injectors.
  • Designed for HPLC systems.
  • Features a 10 μL volume.
  • Compatible with Rheodyne 7010 and 7125 injectors.
  • Supplied as 1 unit per package.
  • Replaced as required for optimal performance.