Learn More
Waters Corp Sample Loop, 10 μL, for Cheminert Valve
Catalog No. 50792761 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Sample loops are integral components within chromatographic injection systems, designed to contain and deliver samples onto a chromatographic column. They work in conjunction with other injection system components, such as the needle and valves, to ensure precise sample aspiration and injection.
  • Designed to contain samples before injection onto a chromatographic column.
  • Accommodates a variety of injection volumes.
  • Functions effectively with full-loop and partial-loop injection modes.
  • Replaceable component for easy system maintenance.
Catalog No. 50-792-761 Supplier Waters Corp Supplier No. 700002504
Sample loops are integral components within chromatographic injection systems, designed to contain and deliver samples onto a chromatographic column. They work in conjunction with other injection system components, such as the needle and valves, to ensure precise sample aspiration and injection.
  • Designed to contain samples before injection onto a chromatographic column.
  • Accommodates a variety of injection volumes.
  • Functions effectively with full-loop and partial-loop injection modes.
  • Replaceable component for easy system maintenance.