Learn More
Trajan Scientific VICI Valco Sample Loop Valco 10l Stainless Steel (VAL.SL10CW)
Catalog No. 50635019
Encompass
VICI Valco 1/16" Stainless Steel Loops Includes Two Stainless Nuts and Two Stainless ferrµles. VICI Valco Part Number: SL10CW (NEW LPP PN: VAL.SL10CW). Thermo Part Number: CH-952458. Made in the USA.
Catalog No. 50-635-019 Supplier Trajan Scientific Supplier No. PAL.SL10CW
VICI Valco 1/16" Stainless Steel Loops Includes Two Stainless Nuts and Two Stainless ferrµles. VICI Valco Part Number: SL10CW (NEW LPP PN: VAL.SL10CW). Thermo Part Number: CH-952458. Made in the USA.