Learn More
Trajan Scientific VICI Valco Sample Loop 1.0-mL Cheminert (VAL.CSL1K)
Catalog No. 50635007
Encompass
VICI Valco Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and ferrµles. VICI Valco Part Number: CSL1K (NEW LPP PN: VAL.CSL1K). Thermo Part Number: CH-953295. Made in the USA.
Catalog No. 50-635-007 Supplier Trajan Scientific Supplier No. PAL.CSL1K
VICI Valco Stainless Steel Loop With 1/16" Tube Ends Includes Two Stainless Steel Nuts and ferrµles. VICI Valco Part Number: CSL1K (NEW LPP PN: VAL.CSL1K). Thermo Part Number: CH-953295. Made in the USA.