Learn More
Waters Corp Sample Loop, 2 mL, Coiled, Stainless Steel
Catalog No. 502391858 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 2 mL coiled sample loop, stainless steel, is the standard sample loop installed in the U6K Injector and used on the Rheodyne injector and Separation Modules.
  • Loop design
  • Compatible with HPLC systems
  • Constructed from stainless steel
  • 2 mL volume
  • Replacement as required for optimal performance
Catalog No. 50-239-1858 Supplier Waters Corp Supplier No. WAT096106
The 2 mL coiled sample loop, stainless steel, is the standard sample loop installed in the U6K Injector and used on the Rheodyne injector and Separation Modules.
  • Loop design
  • Compatible with HPLC systems
  • Constructed from stainless steel
  • 2 mL volume
  • Replacement as required for optimal performance