Learn More
Thermo Scientific Sample Loops for UltiMate WPS-3000T PL RS Pulled Loop Autosamplers | | 1 EA
Supplier:  Thermo Scientific 6826.2450
Encompass
Type:Sample Loop NanoViper, Sample Capacity:1-125 L, Max Pressure:1000 bar, For Use With Equipment:Dionex UltiMate 3000 Pulled Loop Autosampler WPS-3000TPL RS
Catalog No. 50-134-8366