Learn More
Waters Corp Sample Loop, 20 µL
Catalog No. 50792408 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 20 microliter sample loop is one of the smaller sample loops that can be installed to improve the volume repeatability for sample volumes less than 50 microliters..
Catalog No. 50-792-408 Supplier Waters Corp Supplier No. 700000559
The 20 microliter sample loop is one of the smaller sample loops that can be installed to improve the volume repeatability for sample volumes less than 50 microliters..