Learn More
Thermo Scientific Sample Loop 25μL
Supplier:  Thermo Scientific 6850.1911
Encompass
Sample Loop, 25 microliter, biocompatible, for use with Vanquish Split Sampler HT and FT
Catalog No. NC1362963