Learn More
Trajan Scientific VICI Valco Sample Loop, 2mL, For W type Valco Valves, 1/16" Stainless Steel Loop, Includes Two Stainless Nuts and Two Stainless Ferrules (VAL.SL2KCW)
Catalog No. 50635027
Encompass
VICI Valco Sample Loop, 2mL, For W type VICI Valco Valves, 1/16" Stainless Steel Loop, Includes Two Stainless Nuts and Two Stainless Ferrules (NEW LPP PN: VAL.SL2KCW). Made in the USA.
Catalog No. 50-635-027 Supplier Trajan Scientific Supplier No. PAL.SL2KCW
VICI Valco Sample Loop, 2mL, For W type VICI Valco Valves, 1/16" Stainless Steel Loop, Includes Two Stainless Nuts and Two Stainless Ferrules (NEW LPP PN: VAL.SL2KCW). Made in the USA.