Learn More
Waters Corp Waters Corporation Sample Loop, 2 μL, Low Dispersion, ACQUITY UPLC Systems
Catalog No. NC9981333 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 2 μL sample loop is designed for low dispersion applications within UPLC systems. It ensures accurate and reproducible sample introduction onto small-volume UPLC columns to maintain separation efficiency.
  • Designed for low dispersion.
  • Ensures accurate and reproducible sample introduction onto small-volume UPLC columns.
  • Helps maintain separation efficiency.
  • Minimizes sample carryover.
  • Improves peak shape.
  • Maximizes analysis speed.
Catalog No. NC9981333 Supplier Waters Corp Supplier No. 430001264
This 2 μL sample loop is designed for low dispersion applications within UPLC systems. It ensures accurate and reproducible sample introduction onto small-volume UPLC columns to maintain separation efficiency.
  • Designed for low dispersion.
  • Ensures accurate and reproducible sample introduction onto small-volume UPLC columns.
  • Helps maintain separation efficiency.
  • Minimizes sample carryover.
  • Improves peak shape.
  • Maximizes analysis speed.