Learn More
Vici Valco Sample Loop for 1/16" Cheminert HPLC and UHPLC Injectors, 100 µl
SDP
Catalog No. NC1635973
Encompass_Preferred
Metal loops are electrolytically cut and electrochemically polished to ensure square, burr-free ends, then cleaned with microfiltered steam from deionized water. Sample loops are accurately sized for each valve type. Stainless steel loops include stainless nuts and ferrules
Catalog No. NC1635973 Supplier Vici Valco Supplier No. CSL100
Metal loops are electrolytically cut and electrochemically polished to ensure square, burr-free ends, then cleaned with microfiltered steam from deionized water. Sample loops are accurately sized for each valve type. Stainless steel loops include stainless nuts and ferrules