Learn More
Thermo Scientific Sample Loops for UltiMate WPS-3000T PL RS Pulled Loop Autosamplers | | 1 EA
Supplier:  Thermo Scientific 6826.2410
Encompass
Sample loops (1-125µL) for our pulled loop autosampler. Incorporate Thermo Scientific Dionex nanoViper Fingertight Fittings, eliminate dead volumes, ensure easy perfect connections, withstand pressures up to 1000 bar, compatible with third party valves and unions.
Catalog No. 50-134-8363