Learn More
Waters Corp Sample Loop, 500 μL, Stainless Steel, for Model 7125 and 7010
Catalog No. 502390880 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This sample loop is a crucial component for analytical instrumentation, specifically designed to handle precise volumes of samples within chromatographic systems. It ensures accurate and reproducible injections, contributing to reliable experimental results.
  • Capacity of 500 μL for various analytical methods.
  • Compatible with Model 7125 and Model 7010 systems.
  • Constructed from stainless steel, providing durability and chemical resistance.
  • Maintains system performance by ensuring accurate sample delivery.
Catalog No. 50-239-0880 Supplier Waters Corp Supplier No. PSL637026
This sample loop is a crucial component for analytical instrumentation, specifically designed to handle precise volumes of samples within chromatographic systems. It ensures accurate and reproducible injections, contributing to reliable experimental results.
  • Capacity of 500 μL for various analytical methods.
  • Compatible with Model 7125 and Model 7010 systems.
  • Constructed from stainless steel, providing durability and chemical resistance.
  • Maintains system performance by ensuring accurate sample delivery.