Learn More
Waters Corp WATERS CORPORATION Extension Loop, 50 μL, Stainless Steel, HPLC/UPLC
Catalog No. 50466551 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This 50μL sample loop is designed for use with ACQUITY H-Class Sample Manager-FTN and ACQUITY I-Class Sample Manager-FTN modules.
  • Volume: 50 μL
  • Material: Stainless steel
  • Compatible with HPLC and UPLC system types
  • Compatible with Arc Premier System, UPLC H-Class System, UPLC I-Class System, Arc, and APC instruments
  • Includes 1 unit per package
  • Replacement is as required
Catalog No. 50-466-551 Supplier Waters Corp Supplier No. 430002012
This 50μL sample loop is designed for use with ACQUITY H-Class Sample Manager-FTN and ACQUITY I-Class Sample Manager-FTN modules.
  • Volume: 50 μL
  • Material: Stainless steel
  • Compatible with HPLC and UPLC system types
  • Compatible with Arc Premier System, UPLC H-Class System, UPLC I-Class System, Arc, and APC instruments
  • Includes 1 unit per package
  • Replacement is as required